Learn More
Waters Corp Waters Corporation Pressure Relief Valve, 1/4-28 Connection, 35 psi, UPLC & HPLC Systems
Catalog No. 50792742 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement pressure relief valve is designed for use with 2414 and ACQUITY Refractive Index Detectors in UPLC and HPLC systems.
  • 1/4-28 connection
  • 35 psi pressure rating
  • Compatible with 2414 and ACQUITY Refractive Index Detectors
  • Suitable for UPLC and HPLC systems
  • Supplied as 1 unit per package
  • Replacement as required
Catalog No. 50-792-742 Supplier Waters Corp Supplier No. 700002361
This replacement pressure relief valve is designed for use with 2414 and ACQUITY Refractive Index Detectors in UPLC and HPLC systems.
  • 1/4-28 connection
  • 35 psi pressure rating
  • Compatible with 2414 and ACQUITY Refractive Index Detectors
  • Suitable for UPLC and HPLC systems
  • Supplied as 1 unit per package
  • Replacement as required