Learn More
Chemglass Life Sciences Pressure Relief Valve, Complete
SDP
Catalog No. 50873788
Encompass_Preferred
Pressure Relief Valve, Complete. Automatic, adjustable pressure relief valve is for use on apparatus where internal pressure build-up is a concern. Tension on the stainless steel spring is adjusted by turning the top control knob in or out and will relive pressures from 5 to 50 PSI. Two-piece stem is made of PTFE and is fitted with Viton o-rings.
Catalog No. 50-873-788 Supplier Chemglass Life Sciences Supplier No. CG99901
Pressure Relief Valve, Complete. Automatic, adjustable pressure relief valve is for use on apparatus where internal pressure build-up is a concern. Tension on the stainless steel spring is adjusted by turning the top control knob in or out and will relive pressures from 5 to 50 PSI. Two-piece stem is made of PTFE and is fitted with Viton o-rings.