Learn More
PARKER HANNIFIN CORP Single-Use Biopharm Pressure Sensor | 1/2 Barb | 5/pk
SDP
Catalog No. NC1817826
Encompass
This senor can measure and document pressure in a range of filtration, separation and purification applications. The pressure sensor is pre-calibrated and has a unique sensor ID making it NIST traceable and suitable for single-use GMP applications. The pre-calibration and sensor ID information are stored on the sensor's chip. The sensors are validated for exposure to gamma irradiation, autoclave and sodium hydroxide sterilization. It can be used for growth media preparation, bioreactor maintenance, cell culture harvest, buffer preparation, product filtration, tangential flow filtration, virus control, and bulk drug dispense.
Catalog No. NC1817826 Supplier PARKER HANNIFIN CORP Supplier No. 080695PSX3P5
This senor can measure and document pressure in a range of filtration, separation and purification applications. The pressure sensor is pre-calibrated and has a unique sensor ID making it NIST traceable and suitable for single-use GMP applications. The pre-calibration and sensor ID information are stored on the sensor's chip. The sensors are validated for exposure to gamma irradiation, autoclave and sodium hydroxide sterilization. It can be used for growth media preparation, bioreactor maintenance, cell culture harvest, buffer preparation, product filtration, tangential flow filtration, virus control, and bulk drug dispense.