Learn More
BURKLE INC Pressure sprayer Food, 40.58 oz. (1200 ml)
Supplier:  BURKLE INC 03090130
Encompass_Preferred
The Food pressure sprayer is specially designed for direct contact with food and guarantees the highest standards of hygiene.
A major advantage of the pressure sprayer is that it provides a constant, homogeneous spray pattern without constant pumping, even during long work intervals.
The transparent viewing window makes it easy to read the fill level, while the rubberized base ring provides a secure stand. The adjustable nozzle allows you to adjust the spray pattern from a fine mist to a powerful jet. The ergonomically designed handle is comfortable to use.
The quick and easy pressure build-up via the piston rod and the integrated safety valve (max. 3 bar) ensure safety and user-friendliness.
The sprayer can be easily disassembled for thorough and effortless cleaning, ensuring maximum hygiene.
Catalog No. 50-000-25616