Learn More
Caron Products And Services Switch, Pressure, Cut-In Range 3-10 PSI, Cut-Out Range 6-20 PSI, 1/4" NPT
Catalog No. NC2080291
Encompass_Preferred
Switch, Pressure, Cut-In Range 3-10 PSI, Cut-Out Range 6-20 PSI, 1/4" NPT
Catalog No. NC2080291 Supplier Caron Products And Services Supplier No. SWT128
Switch, Pressure, Cut-In Range 3-10 PSI, Cut-Out Range 6-20 PSI, 1/4" NPT