Learn More
Andwin Scientific PRESSURE TRANSDUCER 0-5 VDC
SDP
Catalog No. NC4041161
Encompass
Pressure Transducer, 0-5 VDC output, 24 VDC supply, 0-500 psig, all stainless steel wetted parts, 1/4 NPT male M12 connector 0.4% accuracy Non Cancellable Non Returnable Non Refundable
Catalog No. NC4041161 Supplier Andwin Scientific Supplier No. TD6340
Pressure Transducer, 0-5 VDC output, 24 VDC supply, 0-500 psig, all stainless steel wetted parts, 1/4 NPT male M12 connector 0.4% accuracy Non Cancellable Non Returnable Non Refundable