Learn More
REPLIGEN CORPORATION Repligen Pressure Transducer, Precise Pressure Measurement, Sterile and Calibrated, Pack of 25
SDP
Catalog No. NC3496005
Encompass
The Repligen Pressure Transducer is designed for reliable pressure measurement in bioprocessing, enhancing process control and efficiency. Made from durable polysulfone, it offers corrosion resistance and longevity, and comes calibrated and sterile for critical operations. Its compact size and high accuracy make it ideal for varied applications, ensuring precise performance.
Catalog No. NC3496005 Supplier REPLIGEN CORPORATION Supplier No. ACPM10TCC1S
Add to Cart
Add to Cart
Request a Quote
The Repligen Pressure Transducer is designed for reliable pressure measurement in bioprocessing, enhancing process control and efficiency. Made from durable polysulfone, it offers corrosion resistance and longevity, and comes calibrated and sterile for critical operations. Its compact size and high accuracy make it ideal for varied applications, ensuring precise performance.