Learn More
Ace Glass, Inc. Tube, pressure, 4mL, 150psi, 13mm OD, 10.2cm long, complete with#7 front-seal plug
SDP
Catalog No. NC9335754
Encompass_Preferred
4mL glass pressure tube with #7 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 13mm O.D. x 10.2cm long.
Catalog No. NC9335754 Supplier Ace Glass, Inc. Supplier No. 864812
4mL glass pressure tube with #7 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 13mm O.D. x 10.2cm long.