Learn More
MSC Mi-T-M Hot Water Gas Pressure Washer, 3000 psi, 2.9 gpm, Honda Engine
Catalog No. 502838519
Encompass
This hot water pressure washer effectively removes stubborn oil, grease, and chemicals. It features a direct-drive, crankshaft pump, solid ceramic pistons, and a high- and low-pressure chemical injector. The water output temperature is approximately 190°F, and the water output is 2.9 gpm.
  • Portable with pneumatic tires for easy transportation.
  • Built-in thermal relief valve prevents pump damage.
  • A 270-cc gas engine ensures consistent performance.
  • The Electro Magnetic Firing (EMF) system eliminates the need for a battery, generator, 12V blower motor, or 12V ignition transformer.
  • A top-fired burner assembly enhances fuel efficiency and simplifies servicing.
  • Features a recoil starter.
  • Can burn #1 or #2 diesel fuel or kerosene to heat water.
  • Includes a 36"L spray wand.
Catalog No. 50-283-8519 Supplier MSC Supplier No. 49319528
Add to Cart
Add to Cart
Request a Quote
This hot water pressure washer effectively removes stubborn oil, grease, and chemicals. It features a direct-drive, crankshaft pump, solid ceramic pistons, and a high- and low-pressure chemical injector. The water output temperature is approximately 190°F, and the water output is 2.9 gpm.
  • Portable with pneumatic tires for easy transportation.
  • Built-in thermal relief valve prevents pump damage.
  • A 270-cc gas engine ensures consistent performance.
  • The Electro Magnetic Firing (EMF) system eliminates the need for a battery, generator, 12V blower motor, or 12V ignition transformer.
  • A top-fired burner assembly enhances fuel efficiency and simplifies servicing.
  • Features a recoil starter.
  • Can burn #1 or #2 diesel fuel or kerosene to heat water.
  • Includes a 36"L spray wand.