Learn More
MSC Mi-T-M Cold Water Pressure Washer, 3000 PSI, 3.9 GPM, Diesel, Belt Drive
Catalog No. 502839062
Encompass
This CW Premium Series pressure washer is designed for shipping yards and marine applications. It uses diesel power to remove grime, algae, and scale from boats, docks, and heavy-duty equipment. It is also suitable for oil and mining industry applications.
  • External bypass system protects from heat build-up
  • Forged brass manifold
  • Thermal relief valve
  • Water filter with clear screen for easy inspection
  • Triplex piston pump with ceramic plungers
  • Powder coated steel frame with 1.25-inch tube handles
  • Enclosed belt guard
  • Convenient gun/wand holder and hose wrap
  • 13-inch flat-free tires
  • Quick connect nozzles (0°, 15°, 25°, 40°)
  • 50-foot x 3/8-inch non-marking high-pressure hose
  • Professional-grade insulated trigger gun with safety lock-off
Catalog No. 50-283-9062 Supplier MSC Supplier No. 41383233
Add to Cart
Add to Cart
Request a Quote
This CW Premium Series pressure washer is designed for shipping yards and marine applications. It uses diesel power to remove grime, algae, and scale from boats, docks, and heavy-duty equipment. It is also suitable for oil and mining industry applications.
  • External bypass system protects from heat build-up
  • Forged brass manifold
  • Thermal relief valve
  • Water filter with clear screen for easy inspection
  • Triplex piston pump with ceramic plungers
  • Powder coated steel frame with 1.25-inch tube handles
  • Enclosed belt guard
  • Convenient gun/wand holder and hose wrap
  • 13-inch flat-free tires
  • Quick connect nozzles (0°, 15°, 25°, 40°)
  • 50-foot x 3/8-inch non-marking high-pressure hose
  • Professional-grade insulated trigger gun with safety lock-off