Learn More
MSC Karcher Mobile Hot Water Generator, 5 GPM, 1 ea
Catalog No. 502817636
Encompass
The Karcher Series water heater provides supplemental heat to cold water pressure washers, converting them into hot water units. This electric-powered, diesel-heated device is ETL certified to UL and CSA standards, delivering the benefits of hot water cleaning at a rate of up to 5 gal. per minute.
  • Converts cold water pressure washers to hot water machines
  • Electric-powered and diesel heated operation
  • ETL certified to UL and CSA standards for safety
  • High-efficiency burner produces 380,000 BTU
  • Rugged steel tube frame and ventilated metal cabinet for durability
  • Easy-grip handles and hand truck design for maneuverability
  • Features 10 in tubed pneumatic tires for all-terrain handling
Catalog No. 50-281-7636 Supplier MSC Supplier No. 16617474
The Karcher Series water heater provides supplemental heat to cold water pressure washers, converting them into hot water units. This electric-powered, diesel-heated device is ETL certified to UL and CSA standards, delivering the benefits of hot water cleaning at a rate of up to 5 gal. per minute.
  • Converts cold water pressure washers to hot water machines
  • Electric-powered and diesel heated operation
  • ETL certified to UL and CSA standards for safety
  • High-efficiency burner produces 380,000 BTU
  • Rugged steel tube frame and ventilated metal cabinet for durability
  • Easy-grip handles and hand truck design for maneuverability
  • Features 10 in tubed pneumatic tires for all-terrain handling