This pressurized portable eyewash and body spray unit provides 37 gal. (140 L) of water for emergency irrigation in areas without a continuous water supply. Constructed with an ASME-rated stainless steel tank, the unit is designed for durability and corrosion resistance. It features an integrated 6 ft. drench hose and a pressure-activated flow system for thorough eye and face cleansing, meeting ANSI Z358.1 requirements.
- Tank: 37 gal. (140 L) capacity, ASME-rated stainless steel construction.
- Operational safety: Includes an air pressure gauge, pressure relief valve, and tire-type air fill valve.
- Flow specifications: Provides 0.50 gpm (1.9 L) for eyewash and 4 gpm (15.1 L) for drench hose at operating pressures between 30 psi and 90 psi (2.1 bar to 6.2 bar).
- Components: Features ABS plastic anti-surge heads with dust covers and a chrome-plated brass squeeze valve.
- <