Learn More
Hach Company Pressurized Samp. Relay Module
Supplier:  Hach Company 8520600
Encompass
The pressurized sampling relay module has a high-voltage relay for automatic sample collection from a pressurized sample line The module also has low-voltage connections for optional flow pacing and sampler program control The module can be used with a portable refrigerated or an All-Weather Refrigerated Sampler (AWRS) that has the AS950 controller
Catalog No. 50-002-07216