Learn More
American Research Products Inc PrEST Antigen ADAD2
Catalog No. 5000816750
Encompass
PrEST Antigen ADAD2 antigen APrEST82264 for adenosine deaminase domain containing 2 Recombinant protein fragment of Human ADAD2
Catalog No. 50-008-16750 Supplier American Research Products Inc Supplier No. APREST82264
PrEST Antigen ADAD2 antigen APrEST82264 for adenosine deaminase domain containing 2 Recombinant protein fragment of Human ADAD2