Learn More
American Research Products Inc PrEST Antigen C1orf204
Catalog No. 5000815861
Encompass
PrEST Antigen C1orf204 antigen APrEST84700 for chromosome 1 open reading frame 204 Recombinant protein fragment of Human C1orf204
Catalog No. 50-008-15861 Supplier American Research Products Inc Supplier No. APREST84700
PrEST Antigen C1orf204 antigen APrEST84700 for chromosome 1 open reading frame 204 Recombinant protein fragment of Human C1orf204