Learn More
American Research Products Inc PrEST Antigen GPRIN3
Catalog No. 5000815302
Encompass
PrEST Antigen GPRIN3 antigen APrEST84180 for GPRIN family member 3 Recombinant protein fragment of Human GPRIN3
Catalog No. 50-008-15302 Supplier American Research Products Inc Supplier No. APREST84180
PrEST Antigen GPRIN3 antigen APrEST84180 for GPRIN family member 3 Recombinant protein fragment of Human GPRIN3