Learn More
American Research Products Inc PrEST Antigen HMCES
Catalog No. 5000815389
Encompass
PrEST Antigen HMCES antigen APrEST84170 for 5-hydroxymethylcytosine (hmC) binding ES cell-specific Recombinant protein fragment of Human HMCES
Catalog No. 50-008-15389 Supplier American Research Products Inc Supplier No. APREST84170
PrEST Antigen HMCES antigen APrEST84170 for 5-hydroxymethylcytosine (hmC) binding ES cell-specific Recombinant protein fragment of Human HMCES