Learn More
American Research Products Inc PrEST Antigen PPAN
Catalog No. 5000816536
Encompass
PrEST Antigen PPAN antigen APrEST82713 for peter pan homolog (Drosophila) Recombinant protein fragment of Human PPAN
Catalog No. 50-008-16536 Supplier American Research Products Inc Supplier No. APREST82713
Add to Cart
Add to Cart
PrEST Antigen PPAN antigen APrEST82713 for peter pan homolog (Drosophila) Recombinant protein fragment of Human PPAN