Learn More
American Research Products Inc PrEST Antigen SMCO1
Catalog No. 5000815486
Encompass
PrEST Antigen SMCO1 antigen APrEST84268 for single-pass membrane protein with coiled-coil domains 1 Recombinant protein fragment of Human SMCO1
Catalog No. 50-008-15486 Supplier American Research Products Inc Supplier No. APREST84268
PrEST Antigen SMCO1 antigen APrEST84268 for single-pass membrane protein with coiled-coil domains 1 Recombinant protein fragment of Human SMCO1