Learn More
American Research Products Inc PrEST Antigen SYNDIG1L
Catalog No. 5000816087
Encompass
PrEST Antigen SYNDIG1L antigen APrEST83787 for synapse differentiation inducing 1-like Recombinant protein fragment of Human SYNDIG1L
Catalog No. 50-008-16087 Supplier American Research Products Inc Supplier No. APREST83787
Add to Cart
Add to Cart
PrEST Antigen SYNDIG1L antigen APrEST83787 for synapse differentiation inducing 1-like Recombinant protein fragment of Human SYNDIG1L