Learn More
American Research Products Inc PrEST Antigen TICAM1
Catalog No. 5000816888
Encompass
PrEST Antigen TICAM1 antigen APrEST82629 for toll-like receptor adaptor molecule 1 Recombinant protein fragment of Human TICAM1
Catalog No. 50-008-16888 Supplier American Research Products Inc Supplier No. APREST82629
PrEST Antigen TICAM1 antigen APrEST82629 for toll-like receptor adaptor molecule 1 Recombinant protein fragment of Human TICAM1