Learn More
American Research Products Inc PrEST Antigen TM2D2
Catalog No. 5000815987
Encompass
PrEST Antigen TM2D2 antigen APrEST83701 for TM2 domain containing 2 Recombinant protein fragment of Human TM2D2
Catalog No. 50-008-15987 Supplier American Research Products Inc Supplier No. APREST83701
PrEST Antigen TM2D2 antigen APrEST83701 for TM2 domain containing 2 Recombinant protein fragment of Human TM2D2