Learn More
American Research Products Inc PrEST Antigen TSKS
Catalog No. 5000816812
Encompass
PrEST Antigen TSKS antigen APrEST82608 for testis-specific serine kinase substrate Recombinant protein fragment of Human TSKS
Catalog No. 50-008-16812 Supplier American Research Products Inc Supplier No. APREST82608
PrEST Antigen TSKS antigen APrEST82608 for testis-specific serine kinase substrate Recombinant protein fragment of Human TSKS