Learn More
American Research Products Inc PrEST Antigen ZNF274
Catalog No. 5000816644
Encompass
PrEST Antigen ZNF274 antigen APrEST83539 for zinc finger protein 274 Recombinant protein fragment of Human ZNF274
Catalog No. 50-008-16644 Supplier American Research Products Inc Supplier No. APREST83539
PrEST Antigen ZNF274 antigen APrEST83539 for zinc finger protein 274 Recombinant protein fragment of Human ZNF274