Learn More
American Research Products Inc PrEST Antigen ZNF816
Catalog No. 5000816305
Encompass
PrEST Antigen ZNF816 antigen APrEST82512 for zinc finger protein 816 Recombinant protein fragment of Human ZNF816
Catalog No. 50-008-16305 Supplier American Research Products Inc Supplier No. APREST82512
Add to Cart
Add to Cart
PrEST Antigen ZNF816 antigen APrEST82512 for zinc finger protein 816 Recombinant protein fragment of Human ZNF816