Learn More
Abcam Prestained Protein Ladder - E x tra broad molecular weight (6.5 - 270 kDa), 500UL
Catalog No. 502586569
Encompass_Preferred
Prestained Protein Ladder ab234592 is a three-color protein standard with 10 pre-stained proteins covering a range of MW from 6.5-270 kDa.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-6569 Supplier Abcam Supplier No. AB234592500UL
Prestained Protein Ladder ab234592 is a three-color protein standard with 10 pre-stained proteins covering a range of MW from 6.5-270 kDa.
The product is subject to the following: Abcam Restricted Use Statement