Learn More
Abcam Prestained Protein Ladder - Mid-range molecular weight (10 - 180 kDa), 500UL
Supplier:  Abcam AB234617500UL
Encompass_Preferred
Prestained Protein Ladder ab234617 is a blue protein standard with 11 pre-stained proteins covering a range of MW from 10-180 kDa. ab234617 is designed for monitoring protein seperation in SDS-polyacrylamide gel electrophoresis and verifying Western transfer efficiency on membranes (PVDF, nylon, or nitrocellulose) and for approximating the size of proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-3537