Learn More
Abcam Prestained Protein Ladder - Mid-range molecular weight (10 - 180 kDa), 500UL
Catalog No. 502584841
Encompass_Preferred
10-180 kDa prestained protein ladder is a three-color protein standard with 10 pre-stained proteins. - Suitable for western blot (WB), SDS-PAGE - Designed for monitoring protein separation - Supplied in gel loading buffer and is ready to use
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4841 Supplier Abcam Supplier No. AB116027500UL
10-180 kDa prestained protein ladder is a three-color protein standard with 10 pre-stained proteins. - Suitable for western blot (WB), SDS-PAGE - Designed for monitoring protein separation - Supplied in gel loading buffer and is ready to use
The product is subject to the following: Abcam Restricted Use Statement