Learn More
Abcam Prestained Protein Ladder - Broad molecular weight (10 - 245 kDa), 500UL
Catalog No. NC1721835
Encompass_Preferred
High molecular weight prestained protein ladder 10-245 kDa is a three-color protein standard with 12 prestained proteins. - Cited in over 15 publications - Suitable for western blot (WB), SDS-PAGE - Designed for monitoring protein separation - Supplied in gel loading buffer and is ready to use
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1721835 Supplier Abcam Supplier No. AB116028500UL
High molecular weight prestained protein ladder 10-245 kDa is a three-color protein standard with 12 prestained proteins. - Cited in over 15 publications - Suitable for western blot (WB), SDS-PAGE - Designed for monitoring protein separation - Supplied in gel loading buffer and is ready to use
The product is subject to the following: Abcam Restricted Use Statement