Learn More
LAMDA BIOTECH INC Prestained Protein Marker 11
Catalog No. NC3726400
Encompass_Preferred
The Prestained Protein Marker 11 is a comprehensive molecular weight standard featuring 11 pre-stained recombinant proteins spanning 3-260kDa. This advanced formulation includes eight blue, two red, and one green reference bands for enhanced visualization.
Catalog No. NC3726400 Supplier LAMDA BIOTECH INC Supplier No. PPM11
The Prestained Protein Marker 11 is a comprehensive molecular weight standard featuring 11 pre-stained recombinant proteins spanning 3-260kDa. This advanced formulation includes eight blue, two red, and one green reference bands for enhanced visualization.