Learn More
Proteintech Group Inc Prestained protein marker
SDP
Supplier:  Proteintech Group Inc PL00001500UL
Encompass_Preferred
The PL00001 Regular Range Protein Marker is a ready to use three-color protein standard with 10 prestained proteins covering a wide range of molecular weights from 10 to 180 kDa. The PL00001 Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes, (PVDF, nylon, or nitrocellulose) and for estimating the size of proteins.
Catalog No. 50-225-8721