Learn More
Julabo USA Inc PRESTO A45t Circulator
SDP
Supplier:  Julabo USA Inc 9420452.16.T
Encompass_Preferred
JULABO PRESTO Highly Dynamic Temperature Control Systems for highly precise temperature control of external applications Systems offer rapid heating and cooling rates The hydraulically sealed heat exchanger design affords a wide working temperature range without changing fluids All units feature a color TFT touch screen interface for unit operation external integration and programming Instruments cover a working temperature range of -85 to 250C Up to three user levels with password management Systems incorporate USB (host and device) Ethernet SD-card slot RS232 port stand-by input and Pt100 external temperature sensor port The graphite bearing centrifugal pressure pump is lubricated by the bath fluid All models (except for A30) can operate under pressure control to safeguard pressure sensitive applications (such as jacketed glass reactors) Air and water-cooled versions available Additional accessories available
Catalog No. 50-000-77870
Add to Cart
Request a Quote