Learn More
Imtakt PRESTO FF-C18 HPLC COLUMN
Catalog No. NC0092713
Encompass
This is the world’s first 2u non-porous column. This extremely efficient material works very well for separation of large molecules like polymers, surfactants, protein and even DNA. It has a wide effective analysis range from 5kDa to 50 MDa. 150 x 4.6 mm column
Catalog No. NC0092713 Supplier Imtakt Supplier No. FF005
This is the world’s first 2u non-porous column. This extremely efficient material works very well for separation of large molecules like polymers, surfactants, protein and even DNA. It has a wide effective analysis range from 5kDa to 50 MDa. 150 x 4.6 mm column