Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Equipment) | LabTower TII, Pacific RO, Pacific TII |
| Type | Pretreatment Cartridge |
| Includes | 5 μm Filter w/ Carbon Cartridge, Hardness Stabilizer Cartridge |