Learn More
Mac-Mod Avantor Prevail Organic Acid, 110 A, 3 um, 4.6 x 150 mm HPLC Column
SDP
Catalog No. 501120490
Encompass
vantor Prevail Organic Acid (OA) HPLC columns are silica-based HPLC columns for maximum efficiency and high resolution. Avantor Prevail Organic Acid (OA) HPLC columns can separate common organic acids with a combination of speed, sensitivity and simplicity. A simple acidic phosphate buffer and a Prevail OA column at ambient temperature will separate 11 short chain organic acids in less than 6 minutes. Column selectivity can be readily adjusted by changing pH.
Catalog No. 50-112-0490 Supplier Mac-Mod Supplier No. 88655
vantor Prevail Organic Acid (OA) HPLC columns are silica-based HPLC columns for maximum efficiency and high resolution. Avantor Prevail Organic Acid (OA) HPLC columns can separate common organic acids with a combination of speed, sensitivity and simplicity. A simple acidic phosphate buffer and a Prevail OA column at ambient temperature will separate 11 short chain organic acids in less than 6 minutes. Column selectivity can be readily adjusted by changing pH.