Learn More
Mac-Mod Avantor Prevail C18, 110 A, 5 um, 4.6 x 250 mm HPLC Column
SDP
Catalog No. 501120477
Encompass
Avantor Prevail HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. Avantor Prevail HPLC columns are silica based except for Prevail Carbohydrate ES. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes. Avantor Prevail HPLC columns are stable from highly organic to highly aqueous. Avantor Prevail HPLC columns are specialty phases for specific applications. Avantor Prevail HPLC columns provide excellent sensitivity with microbore and ELSD applications.
Catalog No. 50-112-0477 Supplier Mac-Mod Supplier No. 99210
Avantor Prevail HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. Avantor Prevail HPLC columns are silica based except for Prevail Carbohydrate ES. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes. Avantor Prevail HPLC columns are stable from highly organic to highly aqueous. Avantor Prevail HPLC columns are specialty phases for specific applications. Avantor Prevail HPLC columns provide excellent sensitivity with microbore and ELSD applications.