Learn More
Caron Products And Services PM Kit Model 7001-10 Refrigerated Incubator
Catalog No. NC2182843
Encompass_Preferred
Preventative Maintenance Kit for Model 7001-10 (10 cu. ft. Refrigerated Incubator) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing.  Detailed Instructions Included.
This item is not returnable. View return policy
Catalog No. NC2182843 Supplier Caron Products And Services Supplier No. PM700110
Preventative Maintenance Kit for Model 7001-10 (10 cu. ft. Refrigerated Incubator) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing.  Detailed Instructions Included.