Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Previtamin D3 | 1173-13-3 | 98.7% | 1 MG
SDP

Catalog No. 5000340365
Encompass_Preferred

Previtamin D3 is an intermediate in the production of cholecalciferol (vitamin D3). It is intended for research use only.

  • Intermediate in the production of cholecalciferol (vitamin D3)
  • Intended for research use only
  • Ships with dry ice
  • Store at -80°C
  • Protect from light
  • Store under nitrogen
  • Appearance: Oil
  • Color: Colorless to light yellow

Catalog No. 50-003-40365 Supplier Medchemexpress LLC Supplier No. HY1307051MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Previtamin D3 is an intermediate in the production of cholecalciferol (vitamin D3). It is intended for research use only.

  • Intermediate in the production of cholecalciferol (vitamin D3)
  • Intended for research use only
  • Ships with dry ice
  • Store at -80°C
  • Protect from light
  • Store under nitrogen
  • Appearance: Oil
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.