Learn More
Cayman Chemical PrevItamIn D3 1mg
SDP
Catalog No. 502946196
Encompass_Preferred
An intermediate in the synthesis of vitamin D3; biosynthesis rate is dependent on the solar zenith angle, level of skin pigmentation, and age, among other factors
Catalog No. 50-294-6196 Supplier Cayman Chemical Supplier No. 334091 MG
An intermediate in the synthesis of vitamin D3; biosynthesis rate is dependent on the solar zenith angle, level of skin pigmentation, and age, among other factors