Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prexasertib (dihydrochloride) | 1234015-54-3 | 99.7% | 438.31 | 50 MG
SDP

Catalog No. 5000386906
Encompass_Preferred

Prexasertib dihydrochloride is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor with potent anti-tumor activity. It causes double-stranded DNA breakage and replication catastrophe, leading to apoptosis.

  • Selective CHK1 inhibitor with Ki of 0.9 nM and IC50 of <1 nM
  • Also inhibits CHK2 (IC50=8 nM) and RSK1 (IC50=9 nM)
  • Causes double-stranded DNA breakage
  • Induces replication catastrophe and apoptosis
  • Demonstrates potent anti-tumor activity
  • Inhibits MELK (IC50=38 nM), SIK (IC50=42 nM), BRSK2 (IC50=48 nM), and ARK5 (IC50=64 nM)
  • Requires CDC25A and CDK2 to induce DNA damage
  • Causes chromosomal fragmentation in HeLa cells
  • Induces replication stress and depletes RPA2 available for DNA binding
  • Shows tumor growth inhibition in xenografts

Catalog No. 50-003-86906 Supplier Medchemexpress LLC Supplier No. HY18174A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prexasertib dihydrochloride is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor with potent anti-tumor activity. It causes double-stranded DNA breakage and replication catastrophe, leading to apoptosis.

  • Selective CHK1 inhibitor with Ki of 0.9 nM and IC50 of <1 nM
  • Also inhibits CHK2 (IC50=8 nM) and RSK1 (IC50=9 nM)
  • Causes double-stranded DNA breakage
  • Induces replication catastrophe and apoptosis
  • Demonstrates potent anti-tumor activity
  • Inhibits MELK (IC50=38 nM), SIK (IC50=42 nM), BRSK2 (IC50=48 nM), and ARK5 (IC50=64 nM)
  • Requires CDC25A and CDK2 to induce DNA damage
  • Causes chromosomal fragmentation in HeLa cells
  • Induces replication stress and depletes RPA2 available for DNA binding
  • Shows tumor growth inhibition in xenografts

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.