Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prexasertib dimesylate | 1234015-58-7 | MFCD32693943 | 98.9% | 557.60 | C20H27N7O8S2 | 10 MG
SDP

Catalog No. 5000297925
Encompass_Preferred

Prexasertib dimesylate is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor supplied as a solid research compound. It causes double-stranded DNA breakage and replication catastrophe that lead to apoptosis, and is offered in multiple solid pack sizes and solution formats for laboratory use. Not for human or clinical use.

  • Selective ATP-competitive CHK1 inhibitor.
  • Induces DNA double-strand breaks and replication catastrophe.
  • High purity: 98.9%.
  • Available as solid and as 10 mM solution in DMSO.
  • Solubility data provided for in vitro and in vivo protocols.
  • Recommended storage: 4°C (solid) and -80°C for stock solutions.

Catalog No. 50-002-97925 Supplier Medchemexpress LLC Supplier No. HY18174E10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prexasertib dimesylate is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor supplied as a solid research compound. It causes double-stranded DNA breakage and replication catastrophe that lead to apoptosis, and is offered in multiple solid pack sizes and solution formats for laboratory use. Not for human or clinical use.

  • Selective ATP-competitive CHK1 inhibitor.
  • Induces DNA double-strand breaks and replication catastrophe.
  • High purity: 98.9%.
  • Available as solid and as 10 mM solution in DMSO.
  • Solubility data provided for in vitro and in vivo protocols.
  • Recommended storage: 4°C (solid) and -80°C for stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.