Learn More
Medchemexpress LLC Prexigebersen sodium | 202484-91-1 | 95.6% | 5520.6 (free acid) | C177H206N63Na18O110P17 | 10 MG
SDP
Catalog No. 5000287065
Encompass_Preferred
Prexigebersen sodium is a liposomal-formulated antisense oligonucleotide designed to inhibit synthesis of the adaptor protein Grb2. Supplied as a characterized sodium salt for research use, it is intended for preclinical studies of Grb2-mediated signaling pathways and oncology-related investigations.
  • Antisense oligonucleotide targeting Grb2.
  • Liposomal formulation for enhanced delivery.
  • Sodium salt form for aqueous solubility.
  • High purity suitable for research applications.
  • Available in small-scale research quantities.
  • Includes documentation: datasheet, SDS, COA, and LCMS.
Catalog No. 50-002-87065 Supplier Medchemexpress LLC Supplier No. HY153495A10MG
Prexigebersen sodium is a liposomal-formulated antisense oligonucleotide designed to inhibit synthesis of the adaptor protein Grb2. Supplied as a characterized sodium salt for research use, it is intended for preclinical studies of Grb2-mediated signaling pathways and oncology-related investigations.
  • Antisense oligonucleotide targeting Grb2.
  • Liposomal formulation for enhanced delivery.
  • Sodium salt form for aqueous solubility.
  • High purity suitable for research applications.
  • Available in small-scale research quantities.
  • Includes documentation: datasheet, SDS, COA, and LCMS.