Learn More
Chempoint Com PRF N-HEPTANE 55 GAL DRUM
Supplier:  Chempoint Com 0114370
Encompass
a test fuel component in anti-knock test engines, a 100% heptane fuel is the zero point of the octane rating scale
Catalog No. NC9948616