Learn More
Aviva Systems Biology Corp PRF1 Antibody - N-terminal region (ARP42208_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42208_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRF1. Alternate names: FLH2, HPLH2, MGC65093, P1, PFP, PFN1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-58143