Learn More
Cayman Chemical PRfluoro4methoxybutnoIc A 50mg
SDP
Catalog No. 502917697
Encompass_Preferred
A perfluoroalkyl substance; has been found as a contaminant in wastewater
Catalog No. 50-291-7697 Supplier Cayman Chemical Supplier No. 3726950 MG
A perfluoroalkyl substance; has been found as a contaminant in wastewater