Learn More
Cayman Chemical Perfluorobutylsulfonamide, Purity: ≥95%, CAS: 30334-69-1, 100 mg
SDP
Catalog No. 502915291
Encompass_Preferred
A perfluoroalkyl substance; has been found as a contaminant in wastewater and consumable Tampa Bay oysters
Catalog No. 50-291-5291 Supplier Cayman Chemical Supplier No. 37365100 MG
A perfluoroalkyl substance; has been found as a contaminant in wastewater and consumable Tampa Bay oysters