Learn More
Cayman Chemical Perfluorohexanesulfonic Acid, Purity: ≥95%, CAS: 355-46-4, 100 mg
SDP
Catalog No. 502919345
Encompass_Preferred
A perfluoroalkyl substance; activates PPARα in a reporter assay using COS-1 cells expressing the human or mouse receptor from 5-100 µM; has been found in landfill leachate and sea and river water
Catalog No. 50-291-9345 Supplier Cayman Chemical Supplier No. 37248100 MG
A perfluoroalkyl substance; activates PPARα in a reporter assay using COS-1 cells expressing the human or mouse receptor from 5-100 µM; has been found in landfill leachate and sea and river water