Learn More
Cayman Chemical PRfluorohexnsulfonIc AcId 10mg
SDP
Catalog No. 502915393
Encompass_Preferred
A perfluoroalkyl substance; activates PPARα in a reporter assay using COS-1 cells expressing the human or mouse receptor from 5-100 µM; has been found in landfill leachate and sea and river water
Catalog No. 50-291-5393 Supplier Cayman Chemical Supplier No. 3724810 MG
A perfluoroalkyl substance; activates PPARα in a reporter assay using COS-1 cells expressing the human or mouse receptor from 5-100 µM; has been found in landfill leachate and sea and river water