Learn More
Cayman Chemical Perfluoropropanesulfonic Acid, Purity: ≥90%, CAS: 423-41-6, 100 mg
SDP
Catalog No. 502916285
Encompass_Preferred
A perfluoroalkyl substance; has been found as a contaminant in groundwater in areas treated with aqueous film-forming foam fire extinguishing agents
Catalog No. 50-291-6285 Supplier Cayman Chemical Supplier No. 38156100 MG
A perfluoroalkyl substance; has been found as a contaminant in groundwater in areas treated with aqueous film-forming foam fire extinguishing agents