Learn More
Cayman Chemical PRfluoropropnsulfonIc ACD 10mg
SDP
Catalog No. 502916552
Encompass_Preferred
A perfluoroalkyl substance; has been found as a contaminant in groundwater in areas treated with aqueous film-forming foam fire extinguishing agents
Catalog No. 50-291-6552 Supplier Cayman Chemical Supplier No. 3815610 MG
A perfluoroalkyl substance; has been found as a contaminant in groundwater in areas treated with aqueous film-forming foam fire extinguishing agents